Z544418
Molymod® bonding links
Short bond, translucent
价格与库存信息目前不能提供
packaging
pkg of 25
pkg of 4 pkg
mfr. no.
Molymod
L
1 mm
color
translucent
正在寻找类似产品? Visit 产品对比指南
General description
The bond lengths stated in the table are measured from atom to atom. Bonding links are available in various colors and may be used to represent different bond types. For example, gray links can represent covalent bonds, while purple or white links can represent ionic bonds.
Legal Information
Molymod is a registered trademark of Spiring Enterprises Ltd.