Z555940
Scienceware® Touch Free automatic waste can
plastic, capacity 2.5 gal
价格与库存信息目前不能提供
material
plastic
mfr. no.
Bel-Art
capacity
2.5 gal
正在寻找类似产品? Visit 产品对比指南
General description
Opens and closes automatically, protecting users from contact with the waste receptacle or its contents when disposing of waste of any kind-biological/biohazard materials, paper trash, or recyclables. When a hand or solid object is moved over the top, the Scienceware Touch Free automatic waste can detects the motion and opens the lid to accept waste. It then closes automatically.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Touch Free is a trademark of SP Scienceware, a division of SP
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。