Merck
All Photos(1)

Z558168

Spinner turbines for Varian® spectrometers

room temperature, diam. 5 mm

Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes

manufacturer/tradename

Wilmad STV-5

直径

5 mm

Looking for similar products? Visit Product Comparison Guide

一般描述

Limited VT operation possible of a +40°C about ambient.
Fiberglass turbines are made from high density GFK fiberglass and incorporate a single O-ring imbedded at the turbine top to hold the sample tube firmly into place. These spinners are suitable for Varian spectrometers and are a low cost alternative to full VT versions for sample changers.

法律信息

Varian is a registered trademark of Varian Medical Systems, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service