Z560081
NOWPak® cap connector for 1/8 in. O.D. tubing
fitting size 1/8  in.
NACRES:
NB.01
价格与库存信息目前不能提供
fitting size
1/8  in.
General description
The NOWPak adaptor cap is designed to be used with all NOWPak Bag-in-a-Bottle (BIB) containers to provide sealed chemical dispensing when connected to a pump or vacuum dispense apparatus. Typical uses are direct connection to gradient HPLC and GPC instruments where solvent sparging is not required.
The connector includes a 1/8 in. O.D. (pass through style) tube fitting allowing the chemical feed tube to extend directly to the container bottom. Wetted materials include PTFE, FEP and PFA. The stainless steel filter cup (40 micron) prevents drips during container change-out and keeps tube on container bottom. Ten feet of 1/8 in. tubing is provided. ATMI No. (ND-LD-14).
Legal Information
NOWPAK is a registered trademark of Advanced Technology Materials, Inc.
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。