Z566632
BudgetSensors AFM probe
silicon nitride tip, gold/chromium coating, mode, soft contact
价格与库存信息目前不能提供
material
gold/chromium coating
silicon nitride tip
feature
mode soft contact
mfr. no.
Innovative Solutions, SiNi-30
正在寻找类似产品? Visit 产品对比指南
General description
Useful for surface analysis in materials science, chemistry, physics and biology, and semiconductor applications.
  • Silicon and silicon nitride probes
  • Coatings: magnetic, gold, aluminum reflex, conductive chromium/platinum, gold/chromium
  • Modes: soft, tapping, force modulation, contact