Z566659
BudgetSensors AFM probe
silicon tip, aluminum reflex coating, mode, force modulation
价格与库存信息目前不能提供
material
aluminum reflex coating
silicon tip
feature
mode force modulation
mfr. no.
Innovative Solutions, Multi75Al-10
正在寻找类似产品? Visit 产品对比指南
General description
Useful for surface analysis in materials science, chemistry, physics and biology, and semiconductor applications.
  • Silicon and silicon nitride probes
  • Coatings: magnetic, gold, aluminum reflex, conductive chromium/platinum, gold/chromium
  • Modes: soft, tapping, force modulation, contact