Z602647
Normag dosing funnel
capacity 250 mL
NACRES:
NB.43
价格与库存信息目前不能提供
material
borosilicate glass 3.3
Joint
Joint ST/NS29/32
mfr. no.
Normag
availability
not available in USA and Canada
capacity
250 mL
正在寻找类似产品? Visit 产品对比指南
General description
Jacketed with interchangeable glass needle valve.
Legal Information
Normag is a trademark of Normag Labor- und Prozesstechnik GmbH