Merck
All Photos(1)
Z611530
Julabo® bath test tube racks
to fit 28 test tubes, for open bath circulators, diam. 16-17 mm
NACRES:
NB.77
特点
Holds 28 x 16-17 mm tubes
manufacturer/tradename
Julabo 8970320
参数
+150 °C temp. limit
直径
16-17 mm
Looking for similar products? Visit Product Comparison Guide
法律信息
Julabo is a registered trademark of Julabo USA, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service