Z635421
Swiss Boy lab jacks
load ≤150 kg (with wheels and motor, middle position), platform L × W 400 mm × 400 mm, Model 122MR
价格与库存信息目前不能提供
availability
not available in USA and Canada
parameter
≤150 kg load (with wheels and motor, middle position)
359-629 mm vertical limit
platform L × W
400 mm × 400 mm
color
yellow
正在寻找类似产品? Visit 产品对比指南
General description
Jack can be raised with load in place.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.