Z647977
Best® NightHawk nitrile anti-static gloves
size XL
NACRES:
NB.35
价格与库存信息目前不能提供
material
black gloves
black nitrile
size
XL
sterility
non-sterile
feature
CE compliant
disposable
latex free
powder-free
packaging
pack of 50 ea (boxes of 50)
package of 50 ea
mfr. no.
Best Manufacturing 7700PFTXL
Best Manufacturing, 7700PFTXL
L × thickness
240 mm × 0.10 mm
正在寻找类似产品? Visit 产品对比指南
General description
NightHawk are the latest in disposable nitrile hand protection. Not only is NightHawk an accelerator-free, textured fingertip, powder-free disposable nitrile glove it also offers impressive static-dissipative properties.
These gloves are ideal for any job where static is a problem but hand protection is a necessity.
  • Anti-static
  • Powder-free
  • Accelerator-free
  • Textured fingertips
Legal Information
Best is a registered trademark of Showa Glove Co.