Z655996
Heidolph Reax overhead shakers
REAX 20/4
价格与库存信息目前不能提供
AC/DC input
230 V, 50 Hz
feature
CE compliant
Protection class IP 21
analogue rpm adjustment dial controls
mfr. no.
REAX 20/4
availability
available only in EU
speed
1-16 rpm
W × D × H
490 mm × 465 mm × 520 mm
weight
23 kg
正在寻找类似产品? Visit 产品对比指南
General description
Easy and fast attachment of bottles. Comes complete with attachment for 2L bottles. Vessels of 270mm height and max. diameter of 136mm can be fixed with a tension plate.
Compatibility
for 4 bottles