Z665711
Ohaus® Discoverysemi-micro analytical balances
DV114C, AC/DC input 230 V AC
别名:
analytical scales, weighing balances, laboratory balances
价格与库存信息目前不能提供
material
stainless steel weighing plate
feature
CE compliant
CSA approved
mfr. no.
DV114C
weighing capacity measuring range
110 g, resolution: 0.1 mg
platform diam.
90 mm
正在寻找类似产品? Visit 产品对比指南
General description
Ohaus Discovery series of semi-micro and analytical balances combine unmatched weighing performance with Ohaus′ innovative SmarText software, making it extremely reliable and easy-to-use. The Discovery features durable glass and steel construction and together with advanced AutoCal internal calibration makes it the best performing balance in its class.
Discovery models have capacities upto 310g and are perfect for university research, pharmaceutical research and drug discovery, and general lab applications.
Features and Benefits
  • Highest performance
  • Multiple weighing units
  • Environmental settings
  • Easy to set up and use
  • Advanced AutoCal calibration
  • Four methods of density determination
  • RRS232 interface with full GLP/GMP Protocol
  • Protective in-use cover
  • Up-front level indicator
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。