Z671711

Stuart rocker & roller mixers

Stuart SRT6, AC/DC input 230 V AC (supplied with both UK and EU plugs)

NACRES:
NB.73
价格与库存信息目前不能提供

feature

CE compliant

mfr. no.

Stuart SRT6

parameter

10 kg load
33 rpm speed

正在寻找类似产品? Visit 产品对比指南

General description

These roller mixers provide a gentle, but highly efficient rocking and rolling action.

There are two models available with either 6 or 9 rollers, with fixed speed of 33 rpm.

Designed for easy cleaning with plastic rollers and drip trays made from stainless steel to hold accidental spillages.
Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required. Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C.

  • Rocking and rolling action for complete mixing
  • Six roller design with small footprint
  • Designed for continuous quiet operation
  • Can be used in cold rooms or in incubators
  • Ideal for small vials or bottles (max bottle diam. 135 mm)

Legal Information

Stuart is a trademark of Bibby Sterlin Ltd.
分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。