Z673668
Talboys block heater
AC/DC input 230 V AC, Schuko plug
NACRES:
NB.73
价格与库存信息目前不能提供
material
aluminum plate
mfr. no.
Troemner, 94951X
capacity
, holds 1 block
正在寻找类似产品? Visit 产品对比指南
General description
This multi-purpose unit is ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures.
Economical, versatile and compact. It accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials,microplates, and PCR strips or tubes.
Each block has a thermometer well for measuring block temperature. Heats from ambient to 150°C with two temperature adjustments, low range ambient to 100°C/high range 75°C to 150°C.
The anodized aluminum modular heating blocks provide superior temperature stability and heat transfer.
Heating blocks are sold sperately.
Features and Benefits
  • 紧凑的设计可节省工作台空间
  • 可更换的加热模块,能实现最大的灵活性
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。