Z674443
Duran® DG safety caps
3-ports with stopcock
别名:
dg caps, dg cap
NACRES:
NB.17
价格与库存信息目前不能提供
mfr. no.
DURAN®, 11 684 16
正在寻找类似产品? Visit 产品对比指南
General description
DG safety caps are high-grade PTFE, flexible connection systems which are designed specifically for handling aggressive, gas-forming liquids. Available with 1, 2 or 3 connection ports with stopcocks. Complete with 3.2 mm v-ring seal. GL 45 thread.
  • No dangerous gas emissions
  • No slipped tubes
  • No loss due to evaporation
  • Clean working environment
Legal Information
DURAN is a registered trademark of DWK Life Sciences
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。