Z679194
Reusable laboratory coat
size L
NACRES:
NB.35
价格与库存信息目前不能提供
size
L
feature
CE compliant
availability
not available in USA and Canada
正在寻找类似产品? Visit 产品对比指南
General description
Heavyweight 245 g/m, polyester/cotton material with hydrofoil finish, giving Type 6 limited chemical splash protection.
  • Concealed stud fastening
  • Two hip pockets, one top pocket and side trouser pocket access
  • Finished back length 112 cm with centre back vent
Conforms to prEN1513:1997