Z682721
Ace drying flask, pear-shaped, with side indents,
Joint: ST/NS 24/40, plastic-coated glass
别名:
Aldrich drying flask, pear-shaped, with side indents
NACRES:
NB.43
价格与库存信息目前不能提供
material
pear-shaped flask
plastic-coated glass
single-neck
Joint
Joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
mfr. no.
Ace Glass, 3994-124
Sigma-Aldrich
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC