Z692204
Sharpsbin sharps collection bin
volume 1 L
价格与库存信息目前不能提供
packaging
pkg of 1 ea
volume
1 L
General description
Manufactured to BS7320: 1990 and ISO9002 standards and is UN packing group II approved. All bins have a visiible "Fill" line.