Z692379
3M Organic vapor/dust respirator mask 4251
pkg of 10 ea
价格与库存信息目前不能提供
packaging
pkg of 10 ea
mfr. no.
3M
availability
not available in USA and Canada
General description
Protection against organic gases with a boiling point above 65°C and good warning properties. Complete with re-sealable bag.
Approvals
  • Approved to EN 405:2002
  • Category FFA1P2D, assigned Protection Factor 10 or 1000ppm for Gas and Particulates
  • Suitable for use under COSHH, CAW, CLAW and IRR
  • Exempt from maintenance requirements of CoSHH regulations if disposed of within one month
Legal Information
3M is a trademark of 3M Company