Z692948
Grab Rack
for 16 mm tubes
别名:
culture tube racks, test tube racks, Tube racks
价格与库存信息目前不能提供
material
acrylonitrile-butadiene-styrene (ABS)
feature
Holds 50 x 5-10 mL tubes
packaging
pkg of 5 ea
mfr. no.
Heathrow Scientific, HS24371A
L × W × H
259 nL × 127 mm × 49 mm
正在寻找类似产品? Visit 产品对比指南
General description
Two racks featuring silicon grips to hold tubes in place.
Turn the rack over and the tubes will not fall out! Great for unloading contents while tubes stay in place. Tubes insert and extract with a gentle push or pull. The Plus version also has an eject feature that allows all tubes to all be released simultaneously. ABS rack with silicon griping fingers.
  • No assembly required
  • Not autoclavable
Legal Information
Grab Rack is a trademark of Heathrow Scientific