Z694223
Troemner Lab-Frame standard kit with fiberglass rods
heavy-duty, 75 in. x 72 in.
别名:
laboratory clamps, laboratory support rods
NACRES:
NB.35
价格与库存信息目前不能提供
mfr. no.
Troemner, 916201
frame L × W
75 in. × 72 in.
正在寻找类似产品? Visit 产品对比指南
General description
Standard Lab-Frame kits are available in five sizes and three lattice rod choices:
  • traditional center-less ground aluminum
  • non-corroding fiberglass
  • tough stainless steel
A range of lab-frames specifically designed to fit within laboratory fume hood include stainless steel rods. Nickel-plated connectors and lab-frame feet are standard in all kits. Lab-frame bases may be permanently mounted to bench top with screws (included).
Floor mounted, free-standing unit is specifically designed for heavy-duty work such as pilot plant set ups. Outside frame is constructed of a strong, but light weight steel channel. Interior lattice uses a horizontal channel connector; one end accommodates the rod, the other end fits into the channel and locks tightly with a bolt. Loosen the bolt, and the connector slides easily along the channel track for simple lattice adjustment. Entire apparatus is supported on sturdy, counter-weighted cast iron feet, which may be bolted to the floor for added stability required in top heavy installations. Overall dimensions are 75 in. H (1219 mm) x 72 in. W (1829 mm).
Components:
  • 48 in. (1219 mm) rods (qty 5)
  • 72 in. (1829 mm) rods (qty 3)
  • S-connectors (qty 15)
  • Channel connectors (qty 16)
  • Clamping nuts (qty 24)
  • Cast iron feet, 21.7 in. (qty 2)
  • Frame set (qty 1)
  • Wrench (qty 1)