Merck
Z740685
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 1000 mL
登录to View Organizational & Contract Pricing
物料
borosilicate glass flask
flat bottom flask
接合处
joint stopper10
包装
pkg of 12 ea
容量
1000 mL
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z740686Z740678Z740679
joint
joint stopper10
joint
joint stopper10
joint
joint stopper4
joint
joint stopper4
packaging
pkg of 12 ea
packaging
pkg of 6 ea
packaging
pkg of 12 ea
packaging
pkg of 6 ea
capacity
1000 mL
capacity
1000 mL
capacity
100 mL
capacity
100 mL
一般描述
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
分析证书(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 2
1 of 2
Coors™ Buchner funnel capacity 25 mL
Z247308
Coors Buchner funnel