Z740689
Sigma-Aldrich
Aldrich® Essentials boiling flask, round-bottom
capacity 100 mL, neck diam. 26 mm
NACRES:
NB.43
价格与库存信息目前不能提供
material
borosilicate glass flask
round bottom
single-neck
packaging
pkg of 6 ea
capacity
100 mL
neck diam.
26 mm
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Clear borosilicate glass round bottomed, (Florence), boiling flasks. Narrow long neck and narrow mouth with tooled beaded rim. Large white marking spot and capacity. Designed to meet ASTM E1403 specifications.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC