Merck
All Photos(1)
Z740704
Sigma-Aldrich
Aldrich® Essentials boiling flask, flat-bottom
capacity 5000 mL, neck diam. 50 mm
NACRES:
NB.43
物料
borosilicate glass flask
flat bottom
single-neck
包装
pkg of 1 ea
容量
5000 mL
颈直径
50 mm
Looking for similar products? Visit Product Comparison Guide
一般描述
Clear borosilicate glass flat bottomed boiling flasks. Narrow long neck and narrow mouth with beaded rim. Large white marking spot.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service