Z740755
Sigma-Aldrich
Aldrich® Essentials volumetric flask, class A, meets ASTM E288
capacity 200 mL, tol. 0.150 mL, stopper size size 16, PE
NACRES:
NB.43
价格与库存信息目前不能提供
material
PE
stopper size size
16
Joint
Joint ST/NS neckstopper size 16
feature
class A meets ASTM E288
packaging
pkg of 6 ea
tol. measuring range
0.150 mL
capacity
200 mL
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Clear borosilicate glass volumetric flask with polyethylene stopper. Class A meets ASTM E288 specifications. Blue marking spot and capacity marking. White level marking. Flask are certified and either a batch or individual certificate. Calibrated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC