Z740783
Sigma-Aldrich
Aldrich® Essentials volumetric flask, Class A, meets DIN ISO-1042
capacity 1000 mL, tol. 0.400 mL, stopper size size 22, PE
NACRES:
NB.43
价格与库存信息目前不能提供
material
PE
stopper size size
22
Joint
Joint ST/NS neckstopper size 22
feature
class A meets DIN ISO-1042
packaging
pkg of 6 ea
tol. measuring range
0.400 mL
capacity
1000 mL
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Clear borosilicate glass volumetric flask with polyethylene stopper. Class A, meets DIN ISO-1042 specifications. Blue marking spot and capacity marking. White level marking. Flask are certified and either a batch or individual certificate. Calibrated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC