Z740817
Sigma-Aldrich
Aldrich® Essentials volumetric flask, Class B, meets DIN ISO-1042
capacity 100 mL, tol. 0.20 mL, stopper size size 13, PE
价格与库存信息目前不能提供
material
PE
stopper size size
13
Joint
Joint ST/NS neckstopper size 13
feature
class B meets DIN ISO-1042
packaging
pkg of 6 ea
tol. measuring range
0.20 mL
capacity
100 mL
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Clear borosilicate glass volumetric flask with polyethylene stopper. Class B, Meets DIN ISO-1042 specifications. Blue marking spot and capacity marking. White level marking. Calibrated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC