Z740822
Sigma-Aldrich
Aldrich® Essentials volumetric flask, Class B, meets DIN ISO-1042
capacity 500 mL, tol. 0.50 mL, stopper size size 19, PE
NACRES:
NB.43
价格与库存信息目前不能提供
material
PE
stopper size size
19
Joint
Joint ST/NS neckstopper size 19
feature
class B meets DIN ISO-1042
packaging
pkg of 12 ea
tol. measuring range
0.50 mL
capacity
500 mL
正在寻找类似产品? Visit 产品对比指南
相关类别
General description
Clear borosilicate glass volumetric flask with polyethylene stopper. Class B, Meets DIN ISO-1042 specifications. Blue marking spot and capacity marking. White level marking. Calibrated “To Contain”.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC