Merck
All Photos(1)
Z740951
Sigma-Aldrich
Aldrich® Essentials culture tube with screw cap and PTFE/rubber liner
capacity 50 mL, O.D. × L 25 mm × 150 mm, round bottom
NACRES:
NB.11
物料
borosilicate glass tube
round bottom
screw top cap
screw top neck
transparent tube
特点
neck thread external thread
包装
pack of 192 ea
manufacturer/tradename
Sigma-Aldrich
外径 × 长度
25 mm × 150 mm
容量
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
一般描述
Clear borosilicate glass, round bottom culture tubes with thread top and black phenolic cap. Cap has bonded PTFE faced rubber liner.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service