Merck
All Photos(1)

Z742048

Exact-Temp bottle thermometer with 2 point DAkkS-Calibration

for oven, temperature +35 to +200 °C, accuracy: 1 °C

物料

bottle ( Vermiculite)

描述

OV-080-S

特点

accuracy± 2 °C (>100°C:±3)
calibration (2 point DAkkS)

manufacturer/tradename

70 001 17/04/2P (Ludwig Schneider)

temperature measuring range

+35 to +200 °C, accuracy: 1 °C

长度

180 mm

颜色

red

Looking for similar products? Visit Product Comparison Guide

一般描述

Exact-temp thermometers with DAkkS-Calibration are high-precise instruments to monitor cooling and tempering devices in laboratories, medical technology and R&D.
Each thermometer is mounted in an unbreakable plastic bottle, filled with a special medium to ensure accurate measurings even at open doors.
With DAkkS-Calibration certificate (two calibration points as standard) these high precision thermometers are especially suitable for monitoring certified quality management systems.
They are supplied with magnets for spacesaving wall or door fixing. Exact-temp thermometers are very reliable and economical because they work independent from any mains or battery connection.
Traceable to international standards Exact-temp thermometers are available with organic filling in varying measuring ranges and scale units.
The instruments are carefully aged to ensure a high accuracy. Exact-temp thermometers are calibrated with a reference and are therefore traceable to international standards.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service