Merck
Z742267
DURAN® GL45 thread adapter for bottle top vacuum filtration unit
GL 45 male, GL 56 female, white PTFE
登录to View Organizational & Contract Pricing
Select a Size
物料
white PTFE
manufacturer/tradename
DURAN® 291195601
配件
GL 45 male
GL 56 female
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z742006Z742261S2GPT01RE
DURAN® TILT Media Bottle capacity 500 mL, screw top clear borosilicate glass 3.3 bottle, graduations, non-sterile
Z742261
DURAN® TILT Media Bottle
manufacturer/tradename
DURAN® 291195601
manufacturer/tradename
DURAN® 218812854
manufacturer/tradename
DURAN® 218914459
manufacturer/tradename
-
fitting
GL 45 male, GL 56 female
fitting
-
fitting
-
fitting
inlet funnel, 45 mm outlet thread(s)
一般描述
The re-usable adaptor (GL 45 external/GL 65) allows the use of the DURAN TILT bottle with the 45 mm filtration units for the fitler and sterilization of cell culture media. Manufactured from inert PTFE, can be autoclaved and depyrogenised at 300°C.
法律信息
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 4
1 of 4
DURAN® TILT Light Shield white silicone, includes GL 56 bottle tags
Z742262
DURAN® TILT Light Shield
DURAN® GL 56 Bottle Tag blue silicone
Z742265
DURAN® GL 56 Bottle Tag