Merck
Z742314
2MAG MIX 1 XL single point stirrers up to 40 L
AC/DC input 100-240 V, US 3-pin plug
登录查看公司和协议定价
别名:
high volume stirrer, induction plate, induction stirrer, laboratory stirrer, magnetic stir plate, magnetic stirrer
NACRES:
NB.72
物料
stainless steel housing
stainless steel plate
特点
positions 1
0 ‑ 240 S Variable Intervals (2 step)
Power Setting 5/10/15/20 W ((4-step)
4 step)
包装
pack of 1 ea
制造商/商品名称
2mag 30201
参数
-10-50 °C temp. limit (at 95 percent humidity)
70-2000 rpm speed
宽度 × 直径 × 高度
225 mm × 280 mm × 35 mm
plate size × W × L
× 220 mm × 250 mm
搅拌器体积
1-40 L , per stirring position
重量
~4.0 kg
正在寻找类似产品? Visit 产品对比指南
一般描述
Ultra flat, strong magnetic stirrer with large set-up area and fully automatic stirring bar monitoring mixWatch, up to 40 litres stirring volume, also suitable for highly viscous media, very strong magnetic coupling, therefore predestined for magnetic power transmission over wide distances or for thick vessel bottoms.
ntegrated controller with strong 20 watts stirring power, 4-step power setting for high power with regard to large and viscous stirring volumes and reduced power for the warming-free continuous operation, SoftStart-function (adjustable, 4 steps) for reliable catching/centering and safe accelerating of the stirring bar at different volumes and viscosities, adjustable interval times for automated restart of the stirrer after max. 240 seconds.
分析证书(COA)
已有该产品?
为方便起见,与您过往购买产品相关的文件已保存在文档库中。
访问文档库
难以找到您所需的产品或批次号码?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA