Merck
Z742349
2MAG Asteroid extra strength stirring bar
diam. × L 39 mm × 70 mm
别名:
magentic flea, stirbar, 2Mag 44070, magnetic stirring device, stirring magnet
登录并加购,请在购物车里查看协议价、货期和发货地。
选择规格
变更视图
关于此项目
NACRES:
NB.72
UNSPSC Code:
41103800
Material:
PTFE cover, interior (SmCo), triangular
Diam. × l:
39 mm × 70 mm
Manufacturer/tradename:
2mag 44070
Packaging:
pack of 1 ea
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
material
PTFE cover, interior (SmCo), triangular
packaging
pack of 1 ea
manufacturer/tradename
2mag 44070
diam. × L
39 mm × 70 mm
weight
0.3 kg
General description
New developed, high-effective stirring bar, ideal for large mixing volumes, viscous media, stirring over wide distances or for disadvantageous vessel bottoms.
More than 4x higher torque transmissible, unbeatable increased mixing effect in comparison to conventional stirring bars, high energetic and long-term stable magnetic field, no demagnetization effect, concave design for a minimum of bearing surface and reliable rotation, triangular shape for powerful shear force, better mixing effect at lower rotation speed.
300 percent less PTFE abrasion than comparable round shape stirring bars, significantly higher rotation speed reachable, high quality, even and sealed PTFE cover, FDA-conform, steam sterilizable at 121 degrees C.
Still not finding the right product?
历史批次信息供参考:
分析证书(COA)