Merck
Z742605
myVolt Touch power supply
AC/DC input 230 V AC, EuroPlug
登录to View Organizational & Contract Pricing
Select a Size
AC/DC input
5 - 400 mA (Max Wattage: 80W; Voltage Output: 10-300V (5V increments))
特点
output terminals: 4 pairs
touchscreen control; Dual orientation, auto rotating display
包装
pkg of 1
manufacturer/tradename
Benchmark Scientific E2301-E
相容性
with most brands of gel boxes
正在寻找类似产品? Visit 产品对比指南
一般描述
The Accuris myVolt Touch is a dependable electrophoresis power supply that includes all of the performance and specifications needed for efficient seperations of nucleic acids and proteins. Perfect for agarose gels and protein seperations, the myVolt Touch incorporates some very unique user-friendly features: Touch-screen Control Panel: Setting up an electrophoresis run with the myVolt Touch couldn′t be easier... simply touch the parameter that you want to change and then touch "start" to begin the run.
Two sets of output jacks are available to connect up to 2 gel rigs and the instruments can securely be stacked for use with additional gel rigs.
法律信息
myVolt is a trademark of Benchmark Scientific, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA