Merck
Z742801
Kern EMB series precision balances
Kern EMB 2000-2, weighing capacity 2000 g, accuracy: 0.01 g, precision: ±0.05 g
登录to View Organizational & Contract Pricing
别名:
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
NACRES:
NB.71
manufacturer/tradename
Kern EMB 2000-2
weighing capacity measuring range
2000 g, accuracy: 0.01 g, precision: ±0.05 g
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z742799Z742802Z742794
Kern EMB series precision balances Kern EMB 2000-2, weighing capacity 2000 g, accuracy: 0.01 g, precision: ±0.05 g
Z742801
Kern EMB series precision balances
Kern EMB series precision balances Kern EMB 1000-2, weighing capacity 1000 g, accuracy: 0.01 g, precision: ±0.05 g
Z742799
Kern EMB series precision balances
Kern EMB series precision balances Kern EMB 2200-0, weighing capacity 2200 g, accuracy: 1.0 g, precision: ±2.0 g
Z742802
Kern EMB series precision balances
Kern EMB series precision balances Kern EMB 200-2, weighing capacity 200 g, accuracy: 0.01 g, precision: ±0.02 g
Z742794
Kern EMB series precision balances
weighing capacity measuring range
2000 g, accuracy: 0.01 g, precision: ±0.05 g
weighing capacity measuring range
1000 g, accuracy: 0.01 g, precision: ±0.05 g
weighing capacity measuring range
2200 g, accuracy: 1.0 g, precision: ±2.0 g
weighing capacity measuring range
200 g, accuracy: 0.01 g, precision: ±0.02 g
一般描述
Entry level laboratory balance with tremendous weighing performance
  • Simple and convenient 2-key operation, making them ideal for use in schools and universities
  • Tare function facilitates formulation work
  • Particularly flat design
  • Secure and non-slip positioning with rubber feet
  • Ready for use: Batteries included. AUTO-OFF function to preserve the batteries, can be switched off
分析证书(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA