Merck
All Photos(1)

Z743760

ART® Low Retention SoftFit L Reload System

Pure 200L, size 200 μL, reload insert, sterile

Synonym(s):
aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip
NACRES:
NB.02

无菌性

sterile

特点

Pure 200L
reload insert

包装

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 3751-05-RI

technique(s)

PCR: suitable

尺寸

200 μL

Looking for similar products? Visit Product Comparison Guide

法律信息

ART is a registered trademark of Molecular BioProducts, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service