Merck
All Photos(1)

Z743762

ART® Low Retention SoftFit L Reload System

Pure 20L, size 20 μL, racked, sterile

Synonym(s):
pipet tip, aerosol barrier tips, barrier tip, sterile barrier pipette tip, barrier filter tips, filter pipette tip, filter tip
NACRES:
NB.02

无菌性

sterile

特点

Pure 20L
racked

包装

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 3721-05

technique(s)

PCR: suitable

尺寸

20 μL

Looking for similar products? Visit Product Comparison Guide

法律信息

ART is a registered trademark of Molecular BioProducts, Inc.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service