Z747505
Whatman® FTA® card technology
FTA Micro Card with 1 sample area per card, 125 μL maximum volume/sample area, 125 μL maximum total volume/card, pkg of 25 ea
价格与库存信息目前不能提供
packaging
pkg of 25 ea
mfr. no.
Whatman Article No., 28421894 (US reference)
Whatman, WB120310
正在寻找类似产品? Visit 产品对比指南
Other Notes
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Legal Information
FTA is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。