Z755281

3D Biotek 3D perfusion bioreactor system with pump

12 well chamber with PS scaffold inserts, autoclavable, AC/DC input 240 V AC, European 2-pin plug

NACRES:
NB.22
价格与库存信息目前不能提供

material

polycarbonate (4 interchangeable chambers)

sterility

non-sterile

feature

CE compliant
autoclavable
single use bioreactor system

mfr. no.

3D Biotek

W × D × H

14 in. × 13 in. × 13 in.

matrix active group

non-treated surface treatment

suitability

suitable for (stem cell growth and differentiation)

正在寻找类似产品? Visit 产品对比指南

General description

The 3D perfusion bioreactor consists of multiple independent, autoclavable polycarbonate chambers. The chambers are interchangeable and are specially designed to take advantage of 3D Insert scaffolds with various sizes ranging from 96-well to 6-well. Cell culture medias are perfused through the open porous structure of scaffolds using a pulsatile pump. The entire unit is autoclavable (except pump, of course) and can be used as a single-use bioreactor system.

Application

Numerous studies have proven that cells grow better under dynamic culture condition, simply because both nutrition supply and metabolic waste removal are better under dynamic culture conditions. Additionally, in some instances, the shear force produced by the flowing medium can act as a mechanical stimuli signal that will further promote stem cell differentiation toward certain cell lineage, such as osteoblast. Combined with the 3D Insert scaffold technology, the 3D Perfusion Bioreactor can facilitate the promotion of stem cell growth and differentiation, thereby enhancing stem cell research.

Legal Information

3D Insert is a trademark of 3D Biotek, LLC
分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。