Z767840
EUROSTAR 200 digital overhead stirrer
AC/DC input 230 V AC, UK plug
别名:
laboratory stirrers, electronic overhead stirrers, laboratory mixers
NACRES:
NB.72
价格与库存信息目前不能提供
mfr. no.
IKA, 3990000
parameter
0/6-2000 rpm speed
W × H × D
91 mm × 274 mm × 209 mm
shaft diam. × L
16 mm × 220 mm
stirrer capacity
100 L
output
84 W
正在寻找类似产品? Visit 产品对比指南
General description
Extremely powerful laboratory stirrer for highly viscous applications and intensive mixing for quantities up to 100 L (H2O). It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/6 – 2000 rpm (two speed ranges). Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in viscosities of the sample.
  • Digital speed display
  • Infinitely adjustable speed
  • Push-through agitator shafts
  • Overload protection
  • Short-term overload operation
  • Slim casing
  • Quiet operation
  • Error code display
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。