870852C

Avanti

C18:1(Δ9-cis) Carnitine

oleoyl L-carnitine, chloroform

Empirical Formula (Hill Notation):
C25H47NO4
CAS号:
分子量:
425.64
NACRES:
NA.25
价格与库存信息目前不能提供

packaging

pkg of 1 × 1 mL (870852C-5mg)

mfr. no.

Avanti Polar Lipids, 870852C

concentration

5 mg/mL (870852C-5mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CN+(C)(C)CC@H(OC(CCCCCCC/C=C\CCCCCCCC)=O)CC(O-)=O

InChI

1S/C25H47NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4/h12-13,23H,5-11,14-22H2,1-4H3/b13-12-

InChI key

IPOLTUVFXFHAHI-SEYXRHQNSA-N

Application

C18:1(Δ9-cis) Carnitine may be used as an internal standard in tandem mass spectrometry (MS/MS) for quantitative measurement of carnitines in plasma sample.

Biochem/physiol Actions

C18:1(Δ9-cis) Carnitine, also known as oleoyl L-carnitine, has a potential to inhibit glycine transporter 2 (GLYT2) in a non-competitive manner. It is used as an effective therapeutic for treating pain.

Packaging

5 mL Clear Glass Sealed Ampule (870852C-5mg)

RIDADR

UN 1888 6.1 / PGIII

J E Carland et al.
British journal of pharmacology, 168(4), 891-902 (2012-09-18)
Concentrations of extracellular glycine in the CNS are regulated by two Na(+)/Cl(-) -dependent glycine transporters, GlyT1 and GlyT2. Selective inhibitors of GlyT1 have been developed for the treatment of schizophrenia, whilst selective inhibitors of GlyT2 are analgesic in animal models...
Lawrence C Johnson et al.
Clinical science (London, England : 1979), 132(16), 1765-1777 (2018-06-20)
Advancing age is associated with impairments in numerous physiological systems, leading to an increased risk of chronic disease and disability, and reduced healthspan (the period of high functioning healthy life). The plasma metabolome is thought to reflect changes in the...

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。