N-101

Supelco

Norchlordiazepoxide-D5 solution

100 μg/mL (10% DMSO in Acetonitrile), ampule of 1 mL, certified reference material, Cerilliant®

Empirical Formula (Hill Notation):
C15H7D5ClN3O
分子量:
290.76
NACRES:
NA.24
价格与库存信息目前不能提供

Quality Level

grade

certified reference material

CofA

certificate is enclosed in each package

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

mfr. no.

Cerilliant®

concentration

100 μg/mL (10% DMSO in Acetonitrile)

Featured Industry

Pharmaceutical (small molecule)

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2=[N+]([O-])CC(N)=Nc3ccc(Cl)cc23

正在寻找类似产品? Visit 产品对比指南

General description

This internal standard is suited for quantification of norchlordiazepoxide in LC/MS and GC/MS benzodiazepine testing methods for urine drug testing, forensic analysis, and clinical toxicology applications. Norchlordiazepoxide is a major urinary metabolite of the benzodiazepine chlordiazepoxide, indicated for short term treatment of severe anxiety and for the management of acute alcohol withdrawal syndrome. The metabolite, also known as N-desmethylchlordiazepoxide, is pharmacologically active.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Codes

F,Xn

Risk Statement

11-20/21/22-36

Safety Statement

16-26-36/37

RIDADR

UN 1648 3 / PGII

WGK Germany

2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。