T-082

Supelco

Tiagabine-D6 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Empirical Formula (Hill Notation):
C20H19D6NO2S2 · HCl
CAS号:
分子量:
418.05
EC Number:
NACRES:
NA.24
价格与库存信息目前不能提供

Quality Level

grade

certified reference material

CofA

certificate is enclosed in each package

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

mfr. no.

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

OC([C@@]1([H])CN(CCC=C(C2=C(C([2H])([2H])[2H])C=CS2)C3=C(C([2H])([2H])[2H])C=CS3)CCC1)=O.Cl

InChI

1S/C20H25NO2S2.ClH/c1-14-7-11-24-18(14)17(19-15(2)8-12-25-19)6-4-10-21-9-3-5-16(13-21)20(22)23;/h6-8,11-12,16H,3-5,9-10,13H2,1-2H3,(H,22,23);1H/t16-;/m1./s1/i1D3,2D3;

InChI key

YUKARLAABCGMCN-OUCKJYDLSA-N

General description

A Certified Spiking Solution® suitable for use as an internal standard in LC/MS or GC/MS tiagabine testing methods for clinical toxicology, forensic analysis, urine drug testing, medication monitoring, or pharmaceutical research applications. Tiagabine, marketed as the antiepileptic drug Gabitril®, is used in the treatment of anxiety disorders and neuropathic path including fibromyalgia. The drug has also been approved by US FDA as an adjunctive treatment for partial seizures in ages 12 and up.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Gabitril is a registered trademark of Abbott Laboratories Corp.
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Signal Word

Danger

Target Organs

Eyes

Hazard Codes

F,T

Risk Statement

11-23/24/25-39/23/24/25

Safety Statement

7-16-36/37-45

RIDADR

UN1230 - class 3 - PG 2 - Methanol, solution

WGK Germany

WGK 1

Flash Point(F)

49.5 °F - Not applicable

Flash Point(C)

9.7 °C - Not applicable

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。