T-091

Supelco

2,5-Dimethoxy-4-ethylthiophenethylamine-13C,D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

别名:
2C-T-2-13C,D3 hydrochloride solution
Empirical Formula (Hill Notation):
C1113CH16D3NO2S · HCl
分子量:
281.82
价格与库存信息目前不能提供

Quality Level

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

mfr. no.

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in methanol (as free base)

Featured Industry

Pharmaceutical (small molecule)

storage temp.

−20°C

General description

A stable-labeled internal standard suitable for quantitation of 2C-T-2 levels in urine or other biological matrices by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing. 2C-T-2 is a psychedelic phenethylamine related to the 2C family of recreational drugs such as 2C-T-4, 2C-B, 2C-E, and 2C-I. This designer drug is sold under the street names Blue Mystic and 7th Heaven.

Legal Information

CERILLIANT is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Cerilliant Corporation

Signal Word

Danger

RIDADR

UN1230 - class 3 - PG 2 - Methanol, solution

WGK Germany

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup

分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。