MOULIB384
Sigma-Aldrich
Mouse Oligonucleotide Libraries
(384 well) library
价格与库存信息目前不能提供
storage temp.
−20°C
正在寻找类似产品? Visit 产品对比指南
General description
The Mouse Oligonucleotide Library is a specially designed oligo set that was selected to best represent all of the mouse genes associated with the public mRNA sequences. Using Compugen′s powerful LEADS technology to generate the most comprehensive representation of the transcriptome, a single long oligo has been generated for every gene within the library. These 65-mers have a 5′-C6 amino modification for flexible and universal covalent binding of the oligo to a variety of substrates. The Mouse Oligonucleotide Library consists of 21,997 oligos, which represent 21,766 LEADS clusters plus 231 controls. For more detailed information on the Mouse Oligonucleotide Library, visit www.sigma-genosys.com for the most complete and up-to-date product information.
Features and Benefits
• The most accurate oligos designed by Compugen using their LEADS sequence database and proprietary complex algorithms to provide increased sensitivity and specificity with minimal cross-hybridization
• Libraries are arranged by gene function and are available as an entire library or subsets of the library in 4x96- and 384-well plate format
• Quality tested using PAGE, Mass Spec., and capillary electrophoresis to verify purity, synthesis, length and orientation in plates
• Includes access to
LabOnWeb.com where you will find detailed information about the oligos and genes, including accession numbers, sequence information, oligo properties, gene annotation, and more
Packaging
96-well and 384-well products are both available in 1 nmole/well and 500 pmole/well amounts.
Legal Information
LEADS is a trademark of Compugen
分析证书
原产地证书 (CofO)
联系技术服务部门
社交媒体
LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon
Merck
科研、开发、生产。
作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。
© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。
未经许可,严禁复制本网站上的任何资料。