Merck
All Photos(1)

1.07197

SAFC

胃蛋白酶

(from porcine gastrine mucosa) 0.7 Ph. Eur U/mg, EMPROVE® ESSENTIAL

Synonym(s):
Pepsin
CAS Number:
Molecular Weight:
35000
MDL编号:

Agency

Ph. Eur.

质量水平

500

产品线

EMPROVE® ESSENTIAL

形式

solid

pH值(酸碱度)

4.0-5.0 (20 °C, 10 g/L in H2O)

溶解性

10 g/L

application(s)

pharmaceutical
solid formulation

储存温度

2-8°C

Looking for similar products? Visit Product Comparison Guide

一般描述

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

应用

Pepsin is an endopeptidase that breaks down proteins into smaller peptides. 

包装

5 kg in PE bottle

法律信息

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

象形图

Exclamation markHealth hazard

警示用语:

Danger

危险分类

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

靶器官

Respiratory system

储存分类代码

11 - Combustible Solids

WGK

WGK 1

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Quality

Enter Lot Number to search for Certificate of Quality (COQ).

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials.

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service