Merck
All Photos(1)
1.09208
Millipore
电池组Li-离子
for use with MAS-100 NT®
Agency
non-compendial
特点
electronic
包装
pkg of 1 kit
application(s)
air monitoring
cosmetics
food and beverages
pharmaceutical
相容性
for use with MAS-100 NT®
储存温度
room temp
Related Categories
一般描述
MAS-100 NT®是一种行业标准的空气采样器,旨在监测洁净室和卫生超敏感区域的空气微生物。其高回收率基于Andersen撞击原理。集成气流传感器可补偿环境中压力和温度的差异。 用于MAS-100 NT®的电池组锂离子是一种可充电锂离子电池,其自主性约为5.5小时。
应用
电池组锂离子是MAS-100 NT®的附加配件,可提供长达5.5小时的备用电池。MAS-100 NT® 空气采样器可用于在受控环境(最高ISO 5洁净室)中对环境空气进行微生物监测。微生物空气采样设备广泛用于制药和个人护理行业以及食品、饮料和环境应用中。
包装
1, 1 unit
法律信息
MAS-100 NT is a registered trademark of MBV AG
储存分类代码
10-13 - German Storage Class 10 to 13
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Quality
Enter Lot Number to search for Certificate of Quality (COQ).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service