1.46790

Millipore

Membrane Filter Rinse Fluid

new

Acc. To USP, 6 bottles per box

别名:
Filtration Membrane Rinse Fluid
价格与库存信息目前不能提供

Quality Level

Agency/Method

USP

sterility

sterile; autoclavable

form

liquid

application(s)

microbiological culture: suitable

pH

6.9±0.2 (25 °C)

Featured Industry

Food and Beverages
Pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Membrane Filter Rinse Fluid is designed as a rinsing fluid during membrane filtration process. The balanced nutrients of the fluid protect the retained microbes on the membrane from physiological shock, thus making the microbes capable of cultivating.

Application

Membrane Filter Rinse Fluid is used as a rinsing fluid for filtered samples containing higher hydrocarbons such as Vaseline, paraffin, fats etc. The rinse fluid is used to provide nutrients for the growth of retained microbes on the membrane.

Analysis Note

The appearance of the medium is clear and yellow.
分析证书
原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门

社交媒体

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

Merck

科研、开发、生产。

作为生命科学行业的全球领先供应商,我们致力于为科研、生物技术开发和生产,以及制药药物疗法开发和生产提供各类解决方案和服务。

© 2021年版权归德国达姆施塔特默克集团(Merck KGaA)及/或其附属公司所有。版权所有。

未经许可,严禁复制本网站上的任何资料。