444255

Sigma-Aldrich

MMP Inhibitor Set I - CAS 161314-17-6 - Calbiochem

The MMP Inhibitor Set I controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

别名:
MMP Inhibitor Set I - CAS 161314-17-6 - Calbiochem
CAS号:
MDL编号:

storage conditions

-20C

质量水平

100

形式

solid

manufacturer/tradename

Calbiochem®

储存条件

OK to freeze
desiccated
protect from light

运输

ambient

储存温度

−20°C

一般描述

Contains 5 mg each of MMP-2/MMP-9 Inhibitor I (Cat. No. 444241) and MMP-3 Inhibitor II (Cat. No. 444225), and 1 mg each of GM 6001 (Cat. No. 364205), GM 6001, Negative Control (Cat. No. 364210), and MMP-8 Inhibitor I (Cat. No. 444237).
Matrix metalloproteinases (MMP) are a family of zinc-dependent enzymes that include interstitial collagenases, stromelysins, gelatinases, and membrane-type metalloproteinases. They are part of the intricate extracellular matrix (ECM) and are involved in maintaining the integrity of the ECM by removing undesired proteins. MMPs play a key role in the normal physiology of connective tissue during development, morphogenesis, and wound healing. However, the unregulated activity of MMPs has been implicated in a number of disease processes, including arthritis, tumor cell metastasis, and atherosclerosis. As a result of their involvement in various disease states, MMPs have become a target for the development of potential therapeutic inhibitors.



The Calbiochem brand has assembled a convenient set containing 4 inhibitors (and one negative control) with different specificities for the study of the biological activity of MMPs and the physiological mechanisms of MMP activation and inhibition.

包装

1 set in Glass bottle
Packaged under inert gas

生化/生理作用

Primary Target
MMP

警告

Toxicity: Standard Handling (A)

重悬

Following reconstitution, aliquot and freeze (-20°C).

法律信息

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

分析证书

原产地证书 (CofO)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门