所有图片(1)
963390
Supelco
Adapter with S40 thread with sensor
运输
ambient
相关类别
一般描述
Safe, easy and contamination free solvent handling is one of the main topics in daily lab work.
With this new technology of lab scale level sensoring it is now possible to monitor and manage our bottles in process related steps automatically. If the threshold point of the level detector is dipped into liquid, the termination of the filling process is required by an alarm, and/or an actuator is closed or a pump is switched off. Thus the HPLC-run can be stopped automatically to avoid spilling over of the disposal bottle. The sensor is pre-assembled in the S40-screwcap which is used to connect instruments with 3 mm tubes directly (e.g. HPLC).
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例 T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
请输入1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
Lot and Batch Numbers can be found on a product's label following the words 'Lot' or 'Batch'.
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA